Learn More
Nortriptyline-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Nortriptyline-D3 Hydrochloride |
| CAS | 203784-52-5 |
| Molecular Formula | C19 2H3 H18 N . Cl H |
| Quantity | 0.01 g |
| Synonym | Nortriptyline-D3 Hydrochloride, 3-(10,11-Dihydro-5 H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-1-propanamine hydrochloride, 1-Propanamine, 3-(10,11-dihydro-5 H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-, hydrochloride (1:1), 1-Propanamine, 3-(10,11-dihydro-5 H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-, hydrochloride (9 CI), 3-(10,11-Dihydro-5 H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-1-propanamine hydrochloride (1:1) |
| InChI Formula | InChI=1 S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-11,20 H,6,12-14H2,1H3;1 H/i1D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])NCCC=C1c2ccccc2CCc3ccccc13 |
| IUPAC Name | 3-(5,6-dihydrodibenzo[1,2-a:2',1'-d][7 ]annulen-11-ylidene)-N-(trideuteriomethyl)propan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 302.86 |
| Formula Weight | 302.1629 g/mol |
| Show More |