Learn More
(-)-Norgestrel-2,2,4,6,6,10-d6, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (-)-Norgestrel-d6 |
| CAS | 2376035-98-0 |
| Molecular Formula | C21 D6 H22 O2 |
| Quantity | 0.005 g |
| Synonym | Levonorgestrel-d6, Levonorgestrel-2,2,4,6,6,10-D6, (-)-Norgestrel-2,2,4,6,6,10-d6, 17α-Ethynyl-13β-ethylgon-4-en-17β-ol-3-one-2,2,4,6,6,10-d6, 13-Ethyl-17-hydroxy-18,19-dinor-17α-pregn-4-en-20-yn-3-one-2,2,4,6,6,10-d6 |
| InChI Formula | InChI=1 S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23 H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1/i5D2,6D2,13 D,16 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@@H]3 [C@H](CC[C@@]4(CC)[C@H]3 CC[C@@]4(O)C#C)[C@@]2([2 H])CC([2 H])([2 H])C1=O |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 R)-2,2,4,6,6,10-hexadeuterio-13-ethyl-17-ethynyl-17-hydroxy-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 318.483 |
| Formula Weight | 318.247 g/mol |
| Show More |