Learn More
Norethindrone-2,2,4,6,6,10-d6, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Norethindrone-2,2,4,6,6,10-d6 |
| CAS | 2376036-05-2 |
| Molecular Formula | C20 D6 H20 O2 |
| Quantity | 0.005 g |
| Synonym | Norethisterone D6 (2,2,4,6,6,10-D6), 19-Norpregn-4-en-20-yn-3-one-2,2,4,6,6,10-d6, 17-hydroxy-, (17α)-, (17α)-17-Hydroxy-19-norpregn-4-en-20-yn-3-one-2,2,4,6,6,10-d6, Norethindrone-2,2,4,6,6,10-d6, 4-Estren-17α-ethynyl-17β-ol-3-one-d6, 17α-Ethynyl-19-nortestosterone-d6, 19-Norethisterone-d6, 19-Norethisterone-2,2,4,6,6,10-d6, 19-Norethindrone-2,2,4,6,6,10-d6 |
| InChI Formula | InChI=1 S/C20H26O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,12,15-18,22 H,4-11H2,2H3/t15-,16+,17+,18-,19-,20-/m0/s1/i4D2,5D2,12 D,15 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@@H]3 [C@H](CC[C@@]4(C)[C@H]3 CC[C@@]4(O)C#C)[C@@]2([2 H])CC([2 H])([2 H])C1=O |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 R)-2,2,4,6,6,10-hexadeuterio-17-ethynyl-17-hydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 304.456 |
| Formula Weight | 304.231 g/mol |
| Show More |