missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitenpyram-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Nitenpyram-d3 (N-methyl-d3) |
| Molecular Formula | C11 D3 H12 Cl N4 O2 |
| Quantity | 0.005 g |
| Synonym | Nitenpyram D3 (N-Methyl D3) |
| InChI Formula | InChI=1 S/C11H15ClN4O2/c1-3-15(11(13-2)8-16(17)18)7-9-4-5-10(12)14-6-9/h4-6,8,13 H,3,7H2,1-2H3/b11-8+/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])N\C(=C/[N+](=O)[O-])\N(CC)Cc1ccc(Cl)nc1 |
| IUPAC Name | (E)-1-N'-[(6-chloropyridin-3-yl)methyl]-1-N'-ethyl-2-nitro-1-N-(trideuteriomethyl)ethene-1,1-diamine |
| Molecular Weight (g/mol) | 273.734 |
| Formula Weight | 273.107 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction