Learn More
Nicosulfuron-d6 (dimethoxy-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Nicosulfuron-d6 |
| CAS | 1189419-41-7 |
| Molecular Formula | C15 2H6 H12 N6 O6 S |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C15H18N6O6S/c1-21(2)13(22)9-6-5-7-16-12(9)28(24,25)20-15(23)19-14-17-10(26-3)8-11(18-14)27-4/h5-8 H,1-4H3,(H2,17,18,19,20,23)/i3D3,4D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1cc(OC([2 H])([2 H])[2 H])nc(NC(=O)NS(=O)(=O)c2ncccc2C(=O)N(C)C)n1 |
| IUPAC Name | 2-[[4,6-bis(trideuteriomethoxy)pyrimidin-2-yl]carbamoylsulfamoyl]-N,N-dimethylpyridine-3-carboxamide |
| Molecular Weight (g/mol) | 416.44 |
| Formula Weight | 416.1385 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |