Learn More
(±)-Naproxen-d3 (alpha-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | rac-Naproxen (C-methyl-D3) |
| CAS | 958293-77-1 |
| Molecular Formula | C14 2H3 H11 O3 |
| Quantity | 0.01 g |
| Synonym | 2-Naphthaleneacetic acid, 6-methoxy-α-(methyl-d3)-, rac-Naproxen-D3, rac-Naproxen (C-methyl-D3), [3-D3]-2-(6-Methoxynaphthalen-2-yl)propanoic acid |
| InChI Formula | InChI=1 S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9 H,1-2H3,(H,15,16)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(C(=O)O)c1ccc2cc(OC)ccc2c1 |
| IUPAC Name | 3,3,3-trideuterio-2-(6-methoxynaphthalen-2-yl)propanoic acid |
| Molecular Weight (g/mol) | 233.28 |
| Formula Weight | 233.1131 g/mol |
| Show More |