Learn More
Nabumetone-d3 (methoxy-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Nabumetone-d3 |
| CAS | 1216770-08-9 |
| Molecular Formula | C15 2H3 H13 O2 |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C15H16O2/c1-11(16)3-4-12-5-6-14-10-15(17-2)8-7-13(14)9-12/h5-10 H,3-4H2,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1ccc2cc(CCC(=O)C)ccc2c1 |
| IUPAC Name | 4-[6-(trideuteriomethoxy)naphthalen-2-yl]butan-2-one |
| Molecular Weight (g/mol) | 231.3 |
| Formula Weight | 231.1339 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |