missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Octanoylglycine-2,2-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Octanoylglycine-2,2-D2 |
| CAS | 1219805-51-2 |
| Molecular Formula | C10 D2 H17 N O3 |
| Quantity | 0.01 g |
| Synonym | Glycine-2,2-d2, N-(1-oxooctyl)- (ACI), N-(1-Oxooctyl)glycine-2,2-d2 (ACI), N-Octanoylglycine-2,2-D2, Capryloylglycine-d2, N-Octanoylglycine-2,2-d2 |
| InChI Formula | InChI=1 S/C10H19NO3/c1-2-3-4-5-6-7-9(12)11-8-10(13)14/h2-8H2,1H3,(H,11,12)(H,13,14)/i8D2 |
| SMILES | [2 H]C([2 H])(NC(=O)CCCCCCC)C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-(octanoylamino)acetic acid |
| Molecular Weight (g/mol) | 203.275 |
| Formula Weight | 203.149 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction