missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosodiphenyl-2,2',4,4',6,6'-d6-amine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Nitroso-diphenylamine 2,2',4,4',6,6'-d6 |
| CAS | 93951-95-2 |
| Molecular Formula | C12 2H6 H4 N2 O |
| Quantity | 0.01 g |
| Synonym | Benzen-2,4,6-d3-amine, N-nitroso-N-(phenyl-2,4,6-d3)- (9 CI), N-Nitroso-diphenylamine (2,2',4,4',6,6'-D6), N-Nitroso-bis(2,4,6-trideuteriophenyl)amine |
| InChI Formula | InChI=1 S/C12H10N2O/c15-13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10 H/i1D,2 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1cc([2 H])c(N(N=O)c2c([2 H])cc([2 H])cc2[2 H])c([2 H])c1 |
| IUPAC Name | N,N-bis(2,4,6-trideuteriophenyl)nitrous amide |
| Molecular Weight (g/mol) | 204.26 |
| Formula Weight | 204.117 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction