missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosodi-iso-propyl-d14-amine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Nitrosodiisopropylamine- d14 |
| CAS | 2975202-11-8 |
| Molecular Formula | C6D14N2O |
| Quantity | 0.01 g |
| Synonym | N-(1-Methylethyl)-N-nitroso-2-propanamine- d14, 1,1'-Dimethyl-N-nitrosodiethylamine- d14, Diisopropyl-N-nitrosamine- d14, Diisopropylnitrosamine- d14, Nitrosodiisopropylamine- d14, NSC 336- d14; |
| InChI Formula | InChI=1 S/C6H14N2O/c1-5(2)8(7-9)6(3)4/h5-6 H,1-4H3/i1D3,2D3,3D3,4D3,5 D,6 D |
| SMILES | [2 H]C(N(N=O)C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N-bis(propan-2-yl-d7)nitrous amide |
| Molecular Weight (g/mol) | 144.27 |
| Formula Weight | 144.2 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction