missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-d3-D-proline-2,5,5-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N-Methyl-D-proline-D6 (N-methyl-D3,pyrrolidine-2,5,-D3) |
| Molecular Formula | C6 D6 H5 N O2 |
| Quantity | 0.005 g |
| Synonym | 1-(Methyl-d3)-D-proline-2,5,5-d3, (2 R)-1-(Methyl-d3)-2-pyrrolidinecarboxylic-2,5,5-d3 acid, N-Methyl-D-proline-D6 (N-methyl-D3,pyrrolidine-2,5,-D3) |
| InChI Formula | InChI=1 S/C6H11NO2/c1-7-4-2-3-5(7)6(8)9/h5H,2-4H2,1H3,(H,8,9)/t5-/m1/s1/i1D3,4D2,5 D |
| SMILES | [2 H]C([2 H])([2 H])N1C([2 H])([2 H])CC[C@]1([2 H])C(=O)O |
| IUPAC Name | (2 R)-2,5,5-trideuterio-1-(trideuteriomethyl)pyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 135.194 |
| Formula Weight | 135.117 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction