missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-DL-aspartic-2,3,3-d3 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-D,L-Aspartic Acid 2,3,3-D3 |
| CAS | 89829-69-6 |
| Molecular Formula | C6 D3 H6 N O5 |
| Quantity | 0.01 g |
| Synonym | Aspartic-2,3,3-d3 acid, N-acetyl- (7 CI), 2-Acetamidobutanedioic-2,3,3-d3 acid, N-Acetyl-DL-aspartic-2,3,3-d3 acid, N-Acetyl-DL-aspartic-2,3,3-D3 acid |
| InChI Formula | InChI=1 S/C6H9NO5/c1-3(8)7-4(6(11)12)2-5(9)10/h4H,2H2,1H3,(H,7,8)(H,9,10)(H,11,12)/i2D2,4 D |
| SMILES | [2 H]C([2 H])(C(=O)O)C([2 H])(NC(=O)C)C(=O)O |
| IUPAC Name | 2-acetamido-2,3,3-trideuteriobutanedioic acid |
| Molecular Weight (g/mol) | 178.158 |
| Formula Weight | 178.067 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction