Learn More
N-Acetyl-d3-5-methoxytryptamine, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Melatonin-D3 (N-acetyl-D3) |
| CAS | 90735-69-6 |
| Molecular Formula | C13 D3 H13 N2 O2 |
| Quantity | 0.005 g |
| Synonym | Acetamide-2,2,2-d3, N-[2-(5-methoxy-1 H-indol-3-yl)ethyl]- (9 CI, ACI), N-[2-(5-Methoxy-1 H-indol-3-yl)ethyl]acetamide-2,2,2-d3 (ACI), N-(Acetyl-d3)-5-methoxytryptamine, N-Acetyl-d3-5-methoxytryptamine, Melatonin-d3 |
| InChI Formula | InChI=1 S/C13H16N2O2/c1-9(16)14-6-5-10-8-15-13-4-3-11(17-2)7-12(10)13/h3-4,7-8,15 H,5-6H2,1-2H3,(H,14,16)/i1D3 |
| SMILES | C(CNC(C([2 H])([2 H])[2 H])=O)C=1 C=2 C(NC1)=CC=C(OC)C2 |
| IUPAC Name | 2,2,2-trideuterio-N-[2-(5-methoxy-1 H-indol-3-yl)ethyl]acetamide |
| Molecular Weight (g/mol) | 235.2968 |
| Formula Weight | 235.14 g/mol |
| Show More |