missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mifepristone-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Mifepristone-d3 |
| Molecular Formula | C29 D3 H32 N O2 |
| Quantity | 0.01 g |
| Synonym | Mifepristone D3 (N-methyl D3), Mifepristone-d3, (11β,17β)-11-[4-[(Methyl)(methyl-d3)amino]phenyl]-17-hydroxy-17-(1-propyn-1-yl)estra-4,9-dien-3-one, 17β-Hydroxy-11β-[(methyl)(methyl-d3)amino]phenyl]-17α-(prop-1-ynyl)-estra-4,9-dien-3-one |
| InChI Formula | InChI=1 S/C29H35NO2/c1-5-15-29(32)16-14-26-24-12-8-20-17-22(31)11-13-23(20)27(24)25(18-28(26,29)2)19-6-9-21(10-7-19)30(3)4/h6-7,9-10,17,24-26,32 H,8,11-14,16,18H2,1-4H3/t24-,25+,26-,28-,29-/m0/s1/i3D3 |
| SMILES | [2 H]C([2 H])([2 H])N(C)c1ccc(cc1)[C@H]2 C[C@@]3(C)[C@@H](CC[C@@]3(O)C#CC)[C@@H]4 CCC5=CC(=O)CCC5=C24 |
| IUPAC Name | (8 S,11 R,13 S,14 S,17 S)-17-hydroxy-13-methyl-11-[4-[methyl(trideuteriomethyl)amino]phenyl]-17-prop-1-ynyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 432.6122 |
| Formula Weight | 432.2856 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction