Learn More
Metobromuron-d6 (methoxy-d3; N-methyl-d3), CDN
Product Code. 30318588
Click to view available options
Quantity:
0.005 g
0.01 g
missing translation for 'unitSize'
0.01g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318588
missing translation for 'mfr': CDN CDND78530.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Metobromuron D6 (methyl D3 methoxy D3)
CAS 2732834-84-1
Molecular Formula C9 2H6 H5 Br N2 O2
Quantity 0.005 g
Synonym Metobromuron D6 (methyl D3 methoxy D3)
InChI Formula InChI=1 S/C9H11BrN2O2/c1-12(14-2)9(13)11-8-5-3-7(10)4-6-8/h3-6 H,1-2H3,(H,11,13)/i1D3,2D3
SMILES [2 H]C([2 H])([2 H])ON(C(=O)Nc1ccc(Br)cc1)C([2 H])([2 H])[2 H]
IUPAC Name 3-(4-bromophenyl)-1-(trideuteriomethoxy)-1-(trideuteriomethyl)urea
Molecular Weight (g/mol) 265.14
Formula Weight 264.0381 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity