Learn More
Methyl (2,4-Dichlorophenoxy-d3)acetate, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 2,4-D Methyl Ester-D3 (dichlorophenoxy-D3) |
| CAS | 358731-20-1 |
| Molecular Formula | C9 D3 H5 Cl2 O3 |
| Quantity | 0.01 g |
| Synonym | Acetic acid, (4,6-dichlorophenoxy-2,3,5-d3)-, methyl ester (9 CI), (4,6-Dichlorophenoxy-2,3,5-d3)acetic acid methyl ester, Methyl (2,4-Dichlorophenoxy-d3)acetate, 2,4-D Methyl Ester-d3, 2,4-D Methyl Ester-D3 (dichlorophenoxy-D3) |
| InChI Formula | InChI=1 S/C9H8Cl2O3/c1-13-9(12)5-14-8-3-2-6(10)4-7(8)11/h2-4 H,5H2,1H3/i2D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(OCC(=O)OC)c(Cl)c([2 H])c1Cl |
| IUPAC Name | methyl 2-(2,4-dichloro-3,5,6-trideuteriophenoxy)acetate |
| Molecular Weight (g/mol) | 238.0825 |
| Formula Weight | 237.0039 g/mol |
| Show More |