missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linuron-d6 (methoxy-d3,methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Linuron D6 (methyl D3 methoxy D3) |
| CAS | 1219804-76-8 |
| Molecular Formula | C9 2H6 H4 Cl2 N2 O2 |
| Quantity | 0.005 g |
| Synonym | Linuron D6, 3-(3,4-Dichlorophenyl)-1-(trideuteriomethoxy)-1-(trideuteriomethyl)urea, Linuron-d6, Linuron D6 (methyl D3 methoxy D3) |
| InChI Formula | InChI=1 S/C9H10Cl2N2O2/c1-13(15-2)9(14)12-6-3-4-7(10)8(11)5-6/h3-5 H,1-2H3,(H,12,14)/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])ON(C(=O)Nc1ccc(Cl)c(Cl)c1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 3-(3,4-dichlorophenyl)-1-(trideuteriomethoxy)-1-(trideuteriomethyl)urea |
| Molecular Weight (g/mol) | 255.13 |
| Formula Weight | 254.0496 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction