Learn More
Lansoprazole-d4 (benzimidazole-4,5,6,7-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Lansoprazole-d4 |
| CAS | 934294-22-1 |
| Molecular Formula | C16 2H4 H10 F3 N3 O2 S |
| Quantity | 0.01 g |
| Synonym | 1 H-Benzimidazole-4,5,6,7-d4, 2-[[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfinyl]-, Lansoprazole-D4, Lansoprazole-D4 (benzimidazole-4,5,6,7-D4) |
| InChI Formula | InChI=1 S/C16H14F3N3O2S/c1-10-13(20-7-6-14(10)24-9-16(17,18)19)8-25(23)15-21-11-4-2-3-5-12(11)22-15/h2-7 H,8-9H2,1H3,(H,21,22)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2[nH]c(nc2c1[2 H])S(=O)Cc3nccc(OCC(F)(F)F)c3C |
| IUPAC Name | 4,5,6,7-tetradeuterio-2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1 H-benzimidazole |
| Molecular Weight (g/mol) | 373.39 |
| Formula Weight | 373.101 g/mol |
| Show More |