Learn More
Lamotrigine-d3 (2,3-dichlorophenyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Lamotrigine-D3 |
| CAS | 1132746-94-1 |
| Molecular Formula | C9 D3 H4 Cl2 N5 |
| Quantity | 0.005 g |
| Synonym | 1,2,4-Triazine-3,5-diamine, 6-(5,6-dichlorophenyl-2,3,4-d3)- (ACI), 6-(5,6-Dichlorophenyl-2,3,4-d3)-1,2,4-triazine-3,5-diamine (ACI), Lamotrigine-d3, Lamotrigine-D3, Lamotrigine-d3 (2,3-dichlorophenyl-d3) |
| InChI Formula | InChI=1 S/C9H7Cl2N5/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7/h1-3 H,(H4,12,13,14,16)/i1D,2 D,3 D |
| SMILES | [2 H]c1c([2 H])c(Cl)c(Cl)c(c1[2 H])c2nnc(N)nc2N |
| IUPAC Name | 6-(2,3-dichloro-4,5,6-trideuteriophenyl)-1,2,4-triazine-3,5-diamine |
| Molecular Weight (g/mol) | 259.1099 |
| Formula Weight | 258.0267 g/mol |
| Show More |