missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Homocitrulline-2,6,6-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | L-Homocitrulline-2,6,6-d3 |
| Molecular Formula | C7 D3 H12 N3 O3 |
| Quantity | 0.005 g |
| Synonym | L-Homocitrulline-2,6,6-D3, (2 S)-2-Amino-6-ureidohexanoic-2,6,6-d3 acid, N6-(Aminocarbonyl)-L-lysine-2,6,6-d3 |
| InChI Formula | InChI=1 S/C7H15N3O3/c8-5(6(11)12)3-1-2-4-10-7(9)13/h5H,1-4,8H2,(H,11,12)(H3,9,10,13)/t5-/m0/s1/i4D2,5 D |
| SMILES | [2 H]C([2 H])(CCC[C@]([2 H])(N)C(=O)O)NC(=O)N |
| IUPAC Name | (2 S)-2-amino-6-(carbamoylamino)-2,6,6-trideuteriohexanoic acid |
| Molecular Weight (g/mol) | 192.2308 |
| Formula Weight | 192.1302 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction