missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-2-Pyrrolidinone-3,3,4,4,5-d5-5-carboxylic Acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 5-Oxo-L-proline-d5 |
| CAS | 1086136-22-2 |
| Molecular Formula | C5 D5 H2 N O3 |
| Quantity | 0.01 g |
| Synonym | L-Proline-2,3,3,4,4-d5, 5-oxo- (ACI), 5-Oxo-L-proline-2,3,3,4,4-d5 (ACI), L-Pyroglutamic acid-2,3,4,4,5-d5, L-Pyroglutamic acid-d5, (5 S)-2-Pyrrolidone-5-carboxylic-3,3,4,4,5-d5 acid, Pidolic Acid-D5, L-2-Pyrrolidinone-3,3,4,4,5-d5-5-carboxylic Acid, 5-Oxo-L-proline-d5 |
| InChI Formula | InChI=1 S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1/i1D2,2D2,3 D |
| SMILES | [2 H]C1([2 H])C(=O)N[C@]([2 H])(C(=O)O)C1([2 H])[2 H] |
| IUPAC Name | (2 S)-2,3,3,4,4-pentadeuterio-5-oxopyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 134.1448 |
| Formula Weight | 134.074 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction