missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoproturon-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Isoproturon-d3 |
| CAS | 352438-80-3 |
| Molecular Formula | C12 2H3 H15 N2 O |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C12H18N2O/c1-9(2)10-5-7-11(8-6-10)13-12(15)14(3)4/h5-9 H,1-4H3,(H,13,15)/i3D3 |
| SMILES | [2 H]C([2 H])([2 H])N(C)C(=O)Nc1ccc(cc1)C(C)C |
| IUPAC Name | 1-methyl-3-(4-propan-2-ylphenyl)-1-(trideuteriomethyl)urea |
| Molecular Weight (g/mol) | 209.3 |
| Formula Weight | 209.1607 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction