missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Hyoscyamine-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (-)-Hycosamine-d3 |
| Molecular Formula | C17 D3 H20 N O3 |
| Quantity | 0.005 g |
| Synonym | (-)-Hyoscyamine-d3 (N-methyl-d3), Hyoscyamine-D3 (N-methyl-D3), Hyoscyamine-d3, (αS)-α-(Hydroxymethyl)benzeneacetic acid (3-endo)-8-(methyl-d3)-8-azabicyclo[3.2.1 ]oct-3-yl ester |
| InChI Formula | InChI=1 S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19 H,7-11H2,1H3/t13-,14+,15+,16-/m1/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])N1[C@@H]2 CC[C@H]1 C[C@H](C2)OC(=O)[C@H](CO)c3ccccc3 |
| IUPAC Name | [(1 R,5 S)-8-(trideuteriomethyl)-8-azabicyclo[3.2.1 ]octan-3-yl] (2 S)-3-hydroxy-2-phenylpropanoate |
| Molecular Weight (g/mol) | 292.3879 |
| Formula Weight | 292.1866 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction