missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexestrol-d6 (hexane-2,2,3,4,5,5-d6) (meso), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Hexestrol hexane-2,2,3,4,5,5-d6 |
| CAS | 1219798-48-7 |
| Molecular Formula | C18 2H6 H16 O2 |
| Quantity | 0.005 g |
| Synonym | 4-[2,2,3,4,5,5-Hexadeuterio-4-(4-hydroxyphenyl)hexan-3-yl]phenol, 3,4-Di(p-hydroxyphenyl)-n-hexane-2,2,3,4,5,5-d6, Hexestrol-d6, 3,4-Bis(4-hydroxyphenyl)hexane-2,2,3,4,5,5-d6, Hexestrol-D4 (hexane-2,2,3,4,5,5-D6) |
| InChI Formula | InChI=1 S/C18H22O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,17-20 H,3-4H2,1-2H3/i3D2,4D2,17 D,18 D |
| SMILES | [2 H]C([2 H])(C)C([2 H])(c1ccc(O)cc1)C([2 H])(c2ccc(O)cc2)C([2 H])([2 H])C |
| IUPAC Name | 4-[2,2,3,4,5,5-hexadeuterio-4-(4-hydroxyphenyl)hexan-3-yl]phenol |
| Molecular Weight (g/mol) | 276.4 |
| Formula Weight | 276.1996 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction