missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycolithocholic-2,2,4,4-d4 Acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Glycolithocholic-2,2,4,4-d4 Acid |
| CAS | 2044276-16-4 |
| Molecular Formula | C26 D4 H39 N O4 |
| Quantity | 0.01 g |
| Synonym | Glycine, N-[(3α,5β)-3-hydroxy-24-oxocholan-24-yl-2,2,4,4-d4]-, Glycolithocholic acid (2,2,4,4-D4), N-[(3α,5β)-3-Hydroxy-24-oxocholan-24-yl-2,2,4,4-d4]glycine, 3α-Hydroxy-2,2,4,4-tetradeuterio-5β-cholanoylglycine, 3α-Hydroxy-N-(carboxymethyl)-5β-cholan-24-amide-2,2,4,4-d4, Lithocholylglycine-d4, Lithocholylglycine-2,2,4,4-d4 |
| InChI Formula | InChI=1 S/C26H43NO4/c1-16(4-9-23(29)27-15-24(30)31)20-7-8-21-19-6-5-17-14-18(28)10-12-25(17,2)22(19)11-13-26(20,21)3/h16-22,28 H,4-15H2,1-3H3,(H,27,29)(H,30,31)/t16-,17-,18-,19+,20-,21+,22+,25+,26-/m1/s1/i10D2,14D2 |
| SMILES | [2 H]C1([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3 CC[C@@H]2 C([2 H])([2 H])[C@@H]1 O)[C@H](C)CCC(=O)NCC(=O)O |
| IUPAC Name | 2-[[(4 R)-4-[(3 R,5 R,8 R,9 S,10 S,13 R,14 S,17 R)-2,2,4,4-tetradeuterio-3-hydroxy-10,13-dimethyl-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid |
| Molecular Weight (g/mol) | 437.6486 |
| Formula Weight | 437.3443 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction