missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glipizide-d11 (cyclohexyl-d11), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Glipizide-d11 |
| CAS | 1189426-07-0 |
| Molecular Formula | C21 D11 H16 N5 O4 S |
| Quantity | 0.01 g |
| Synonym | 2-Pyrazinecarboxamide, N-[2-[4-[[[(cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl- (ACI), N-[2-[4-[[[(Cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-amino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl-2-pyrazinecarboxamide (ACI), Glipizide D11 (cyclohexyl D11) |
| InChI Formula | InChI=1 S/C21H27N5O4S/c1-15-13-24-19(14-23-15)20(27)22-12-11-16-7-9-18(10-8-16)31(29,30)26-21(28)25-17-5-3-2-4-6-17/h7-10,13-14,17 H,2-6,11-12H2,1H3,(H,22,27)(H2,25,26,28)/i2D2,3D2,4D2,5D2,6D2,17 D |
| SMILES | [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(NC(=O)NS(=O)(=O)c2ccc(CCNC(=O)c3cnc(C)cn3)cc2)C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 5-methyl-N-[2-[4-[(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)carbamoylsulfamoyl]phenyl]ethyl]pyrazine-2-carboxamide |
| Molecular Weight (g/mol) | 456.603 |
| Formula Weight | 456.247 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction