Learn More
Fluvoxamine-d4 Maleate (aminoethyl-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (E)-Fluvoxamine-d4 Maleate |
| CAS | 1432075-74-5 |
| Molecular Formula | C15 D4 H17 F3 N2 O2 . C4 H4 O4 |
| Quantity | 0.01 g |
| Synonym | 5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone (E)-O-(2-aminoethyl-1,1,2,2-d4)oxime (2 Z)-2-butenedioate, 2-[[[(1 E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxy]ethanamine-1,1,2,2-d4 maleate, (E)-Fluvoxamine-d4 maleate, Fluvoxamine-D4 (aminoethyl-D4) maleate |
| InChI Formula | InChI=1 S/C15H21F3N2O2.C4H4O4/c1-21-10-3-2-4-14(20-22-11-9-19)12-5-7-13(8-6-12)15(16,17)18;5-3(6)1-2-4(7)8/h5-8 H,2-4,9-11,19H2,1H3;1-2 H,(H,5,6)(H,7,8)/b20-14+;2-1-/i9D2,11D2; |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])O\N=C(/CCCCOC)\c1ccc(cc1)C(F)(F)F.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;1,1,2,2-tetradeuterio-2-[(E)-[5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxyethanamine |
| Molecular Weight (g/mol) | 438.4315 |
| Formula Weight | 438.1916 g/mol |
| Show More |