missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Fluoxetine-d5 Oxalate (phenyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Fluoxetine-D5 (phenyl-D5) Oxalate |
| CAS | 1219804-82-6 |
| Molecular Formula | C17 D5 H13 F3 N O . C2 H2 O4 |
| Quantity | 0.01 g |
| Synonym | (±)-Fluoxetine-d5 Oxalate (phenyl-d5), Fluoxetine-D5 (phenyl-D5) Oxalate, Fluoxetine-d5 oxalate, N-Methyl-3-(2,3,4,5,6-pentadeuteriophenyl)-3-[4-(trifluoromethyl)phenoxy]propan-1-amine ethanedioate (1:1) |
| InChI Formula | InChI=1 S/C17H18F3NO.C2H2O4/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;3-1(4)2(5)6/h2-10,16,21 H,11-12H2,1H3;(H,3,4)(H,5,6)/i2D,3 D,4 D,5 D,6 D; |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(CCNC)Oc2ccc(cc2)C(F)(F)F)c([2 H])c1[2 H].OC(=O)C(=O)O |
| IUPAC Name | N-methyl-3-(2,3,4,5,6-pentadeuteriophenyl)-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;oxalic acid |
| Molecular Weight (g/mol) | 404.3918 |
| Formula Weight | 404.1607 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction