missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Fenbuconazole-d5 (phenyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Fenbuconazole (phenyl D5) |
| CAS | 1398066-06-2 |
| Molecular Formula | C19 2H5 H12 Cl N4 |
| Quantity | 0.005 g |
| Synonym | Fenbuconazole (phenyl D5) |
| InChI Formula | InChI=1 S/C19H17ClN4/c20-18-8-6-16(7-9-18)10-11-19(12-21,13-24-15-22-14-23-24)17-4-2-1-3-5-17/h1-9,14-15 H,10-11,13H2/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C(CCc2ccc(Cl)cc2)(Cn3cncn3)C#N |
| IUPAC Name | 4-(4-chlorophenyl)-2-(2,3,4,5,6-pentadeuteriophenyl)-2-(1,2,4-triazol-1-ylmethyl)butanenitrile |
| Molecular Weight (g/mol) | 341.85 |
| Formula Weight | 341.1456 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction