Learn More
Enalapril-d5 Maleate (phenyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Enalapril-d5 Maleate Salt |
| CAS | 349554-02-5 |
| Molecular Formula | C20 D5 H23 N2 O5 . C4 H4 O4 |
| Quantity | 0.01 g |
| Synonym | L-Proline, N-[1-(ethoxycarbonyl)-3-(phenyl-d5)propyl]-L-alanyl-, (2 Z)-2-butenedioate (1:1) (9 CI), N-[(1 S)-1-(Ethoxycarbonyl)-3-(phenyl-d5)propyl]-L-alanyl-L-proline maleate, Enalapril-d5 maleate, Enalapril-D5 (phenyl-D5) maleate |
| InChI Formula | InChI=1 S/C20H28N2O5.C4H4O4/c1-3-27-20(26)16(12-11-15-8-5-4-6-9-15)21-14(2)18(23)22-13-7-10-17(22)19(24)25;5-3(6)1-2-4(7)8/h4-6,8-9,14,16-17,21 H,3,7,10-13H2,1-2H3,(H,24,25);1-2 H,(H,5,6)(H,7,8)/b;2-1-/t14-,16-,17-;/m0./s1/i4D,5 D,6 D,8 D,9 D; |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CC[C@H](N[C@@H](C)C(=O)N2CCC[C@H]2 C(=O)O)C(=O)OCC)c([2 H])c1[2 H].OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;(2 S)-1-[(2 S)-2-[[(2 S)-1-ethoxy-1-oxo-4-(2,3,4,5,6-pentadeuteriophenyl)butan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 497.55 |
| Formula Weight | 497.242 g/mol |
| Show More |