missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Donepezil-d4 HCl (bismethylene-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Donepezil-D4 (1-(phenylmethyl-D2)-4-piperidinyl]methyl-D2) Hydrochloride |
| CAS | 1219798-88-5 |
| Molecular Formula | C24 D4 H25 N O3 . Cl H |
| Quantity | 0.01 g |
| Synonym | 2,3-Dihydro-5,6-dimethoxy-2-[[1-(phenylmethyl-d2)-4-piperidinyl]methyl-d2]-1 H-inden-1-one hydrochloride, Donepezil-d4 hydrochloride, Donepezil-D4 (1-(phenylmethyl-D2)-4-piperidinyl]methyl-D2) hydrochloride |
| InChI Formula | InChI=1 S/C24H29NO3.ClH/c1-27-22-14-19-13-20(24(26)21(19)15-23(22)28-2)12-17-8-10-25(11-9-17)16-18-6-4-3-5-7-18;/h3-7,14-15,17,20 H,8-13,16H2,1-2H3;1 H/i12D2,16D2; |
| SMILES | Cl.[2 H]C([2 H])(C1CCN(CC1)C([2 H])([2 H])c2ccccc2)C3Cc4cc(OC)c(OC)cc4C3=O |
| IUPAC Name | 2-[dideuterio-[1-[dideuterio(phenyl)methyl]piperidin-4-yl]methyl]-5,6-dimethoxy-2,3-dihydroinden-1-one;hydrochloride |
| Molecular Weight (g/mol) | 419.9776 |
| Formula Weight | 419.2165 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction