Learn More
DL-Histidine-alpha,beta,beta-d3, CDN
Product Code. 30318970
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318970
Brand: CDN CDND22720.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DL-Histidine-alpha,beta,beta-d3
CAS 344299-50-9
Molecular Formula C6 D3 H6 N3 O2
Quantity 0.005 g
Synonym DL-Histidine-α,β,β-D3, Histidine-α,β,β-d3 (9 CI)
InChI Formula InChI=1 S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5 H,1,7H2,(H,8,9)(H,10,11)/i1D2,5 D
SMILES [2 H]C([2 H])(c1c[nH]cn1)C([2 H])(N)C(=O)O
IUPAC Name 2-amino-2,3,3-trideuterio-3-(1 H-imidazol-4-yl)propanoic acid
Molecular Weight (g/mol) 158.18
Formula Weight 158.0883 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity