missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Glufosinate-d8 HCl (2,3,3,4,4-d5; methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Glufosinate Hydrochloride D8 (Methyl D3, 2,3,3,4,4-D5) |
| Molecular Formula | C5 D8 H4 N O4 P . Cl H |
| Quantity | 0.005 g |
| Synonym | Glufosinate Hydrochloride D8 (Methyl D3, 2,3,3,4,4 D5), DL-Glufosinate-d8 HCl (2,3,3,4,4-d5, methyl-d3), 2-Amino-4-[hydroxy(methyl-d3)phosphinyl]butanoic2,3,3,4,4-d5 acid hydrochloride |
| InChI Formula | InChI=1 S/C5H12NO4P.ClH/c1-11(9,10)3-2-4(6)5(7)8;/h4H,2-3,6H2,1H3,(H,7,8)(H,9,10);1 H/i1D3,2D2,3D2,4 D; |
| SMILES | Cl.[2 H]C([2 H])([2 H])P(=O)(O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])(N)C(=O)O |
| IUPAC Name | 2-amino-2,3,3,4,4-pentadeuterio-4-[hydroxy(trideuteriomethyl)phosphoryl]butanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 225.6371 |
| Formula Weight | 225.0773 g/mol |
| Percent Purity | 98 atom % D, min 96% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction