missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2,4-Diaminobutyric-2,3,3,4,4-d5 Acid 2 HCl, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | DL-2,4-Diaminobutyric Acid-2,3,3,4,4-D5 Dihydrochloride |
| Molecular Formula | C4 D5 H5 N2 O2 . 2 Cl H |
| Quantity | 0.01 g |
| Synonym | 2,4-Diamino-2,3,3,4,4-pentadeuteriobutanoic acid dihydrochloride, 2,4-Diaminobutanoic acid-d5 dihydrochloride, 2,4-Diaminobutanoic-2,3,3,4,4-d5 acid dihydrochloride, 2,4-Diaminobutyric acid-2,3,3,4,4-d5 dihydrochloride, DL-2,4-Diaminobutyric acid-2,3,3,4,4-D5 dihydrochloride, DL-DABA-d5 dihydrochloride, DL-2,4-Diaminobutyric-2,3,3,4,4-d5 Acid 2 HCl |
| InChI Formula | InChI=1 S/C4H10N2O2.2 ClH/c5-2-1-3(6)4(7)8;;/h3H,1-2,5-6H2,(H,7,8);2*1 H/i1D2,2D2,3 D;; |
| SMILES | Cl.Cl.[2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])(N)C(=O)O |
| IUPAC Name | 2,4-diamino-2,3,3,4,4-pentadeuteriobutanoic acid;dihydrochloride |
| Molecular Weight (g/mol) | 196.0871 |
| Formula Weight | 195.059 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction