Learn More
Dimethyl-d6 Tetrachloroterephthalate, CDN
Product Code. 30321884
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321884
Brand: CDN CDND50840.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chlorthal-dimethyl-d6
CAS 350818-66-5
Molecular Formula C10 D6 Cl4 O4
Quantity 0.01 g
Synonym Dimethyl-d6 Tetrachloroterephthalate
InChI Formula InChI=1 S/C10H6Cl4O4/c1-17-9(15)3-5(11)7(13)4(10(16)18-2)8(14)6(3)12/h1-2H3/i1D3,2D3
SMILES [2 H]C([2 H])([2 H])OC(=O)c1c(Cl)c(Cl)c(C(=O)OC([2 H])([2 H])[2 H])c(Cl)c1Cl
IUPAC Name bis(trideuteriomethyl) 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate
Molecular Weight (g/mol) 338
Formula Weight 335.9397 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity