Learn More
Desipramine-2,4,6,8-d4 HCl, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Desipramine-2,4,6,8-d4 Hydrochloride |
| CAS | 61361-34-0 |
| Molecular Formula | C18 2H4 H18 N2 . Cl H |
| Quantity | 0.005 g |
| Synonym | 5 H-Dibenz[b,f]azepine-2,4,6,8-d4-5-propanamine, 10,11-dihydro-N-methyl-, monohydrochloride (9 CI), Desipramine-2,4,6,8-D4 Hydrochloride |
| InChI Formula | InChI=1 S/C18H22N2.ClH/c1-19-13-6-14-20-17-9-4-2-7-15(17)11-12-16-8-3-5-10-18(16)20;/h2-5,7-10,19 H,6,11-14H2,1H3;1 H/i2D,3 D,9 D,10 D; |
| SMILES | Cl.[2 H]c1cc([2 H])c2N(CCCNC)c3c([2 H])cc([2 H])cc3CCc2c1 |
| IUPAC Name | N-methyl-3-(1,3,8,10-tetradeuterio-5,6-dihydrobenzo[b][1 ]benzazepin-11-yl)propan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 306.87 |
| Formula Weight | 306.1801 g/mol |
| Show More |