missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Denatonium-d5 Benzoate (benzyl-2,3,4,5,6-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Denatonium-D5 (benzyl-2,3,4,5,6-D5) Benzoate |
| Molecular Formula | C21 D5 H24 N2 O . C7 H5 O2 |
| Quantity | 0.01 g |
| Synonym | (Benzyl-2,3,4,5,6-d5)diethyl[(2,6-xylylcarbamoyl)methyl]ammonium benzoate, N-[2-[(2,6-Dimethylphenyl)amino]-2-oxoethyl]-N,N-diethylbenzene-d5-methanaminium benzoate, Denatonium-d5 benzoate, Denatonium-D5 (benzyl-2,3,4,5,6-D5) benzoate, Bitrex-d5 |
| InChI Formula | InChI=1 S/C21H28N2O.C7H6O2/c1-5-23(6-2,15-19-13-8-7-9-14-19)16-20(24)22-21-17(3)11-10-12-18(21)4;8-7(9)6-4-2-1-3-5-6/h7-14 H,5-6,15-16H2,1-4H3;1-5 H,(H,8,9)/i7D,8 D,9 D,13 D,14 D; |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C[N+](CC)(CC)CC(=O)Nc2c(C)cccc2C)c([2 H])c1[2 H].[O-]C(=O)c3ccccc3 |
| IUPAC Name | [2-(2,6-dimethylanilino)-2-oxoethyl]-diethyl-[(2,3,4,5,6-pentadeuteriophenyl)methyl]azanium;benzoate |
| Molecular Weight (g/mol) | 451.612 |
| Formula Weight | 451.2883 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction