Learn More
Daidzein-d4 (4-hydroxyphenyl-2,3,5,6-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Daidzein-2',3',5',6'-D4 |
| CAS | 1219803-57-2 |
| Molecular Formula | C15 D4 H6 O4 |
| Quantity | 0.005 g |
| Synonym | Daidzein-d4 (4-hydroxyphenyl-2,3,5,6-d4), 7-Hydroxy-3-(4-hydroxyphenyl-2,3,5,6-d4)-4 H-1-benzopyran-4-one, Daidzein-2',3',5',6'-d4, Daidzein-d4, Daidzein-2',3',5',6'-D4 |
| InChI Formula | InChI=1 S/C15H10O4/c16-10-3-1-9(2-4-10)13-8-19-14-7-11(17)5-6-12(14)15(13)18/h1-8,16-17 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(C2=COc3cc(O)ccc3C2=O)c([2 H])c([2 H])c1O |
| IUPAC Name | 7-hydroxy-3-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)chromen-4-one |
| Molecular Weight (g/mol) | 258.2622 |
| Formula Weight | 258.083 g/mol |
| Show More |