missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Gluconic-2,3,4,5,6,6-d6 Acid Sodium Salt, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sodium D-Gluconate-2,3,4,5,6,6-D6 |
| Molecular Formula | C6 D6 H5 O7 . Na |
| Quantity | 0.005 g |
| Synonym | D-Gluconic-2,3,4,5,6,6-d6 Acid Sodium Salt, D-Gluconic Acid-2,3,4,5,6,6-d6 Sodium Salt, Sodium D-Gluconate-2,3,4,5,6,6-d6, Sodium D-Gluconate-2,3,4,5,6,6-D6 |
| InChI Formula | InChI=1 S/C6H12O7.Na/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11 H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1/i1D2,2 D,3 D,4 D,5 D; |
| SMILES | [Na+].[2 H]C([2 H])(O)[C@@]([2 H])(O)[C@@]([2 H])(O)[C@]([2 H])(O)[C@@]([2 H])(O)C(=O)[O-] |
| IUPAC Name | sodium;(2 R,3 S,4 R,5 R)-2,3,4,5,6,6-hexadeuterio-2,3,4,5,6-pentahydroxyhexanoate |
| Molecular Weight (g/mol) | 224.1741 |
| Formula Weight | 224.0779 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction