Learn More
D-alpha-Phenyl-d5-glycine, CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | D-(-)-2-Phenylglycine-d5 |
| CAS | 1246817-98-0 |
| Molecular Formula | C8 D5 H4 N O2 |
| Quantity | 0.01 g |
| Synonym | (2 R)-2-Amino-2-(2,3,4,5,6-pentadeuteriophenyl)acetic acid, (2 R)-2-Amino-2-(phenyl-d5)acetic acid, D-Phenylglycine-d5, (αR)-α-Aminobenzene-d5-acetic acid, D-Phenylglycine-D5 (phenyl-D5) |
| InChI Formula | InChI=1 S/C8H9NO2/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7 H,9H2,(H,10,11)/t7-/m1/s1/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c([C@@H](N)C(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | (2 R)-2-amino-2-(2,3,4,5,6-pentadeuteriophenyl)acetic acid |
| Molecular Weight (g/mol) | 156.193 |
| Formula Weight | 156.095 g/mol |
| Show More |