missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-alpha-(4-Hydroxyphenyl-2,3,5,6-d4)glycine-alpha-d1, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | D-a-(4-Hydroxyphenyl-2,3,5,6-d4)glycine-a-d1 |
| Molecular Formula | C8 D5 H4 N O3 |
| Quantity | 0.005 g |
| Synonym | (2 R)-2-Amino-2-deuterio-2-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetic acid, D-(-)-4-Hydroxyphenyl-d4-glycine-d, D-4-Hydroxyphenylglycine-d5, D-(4-Hydroxyphenyl-d4)glycine-α-d, (2 R)-2-Amino-2-(4-hydroxyphenyl-d4)acetic-d acid, (2 R)-2-Amino-2-(4-hydroxyphenyl-2,3,5,6-d4)acetic-2-d acid, (2 R)-2-Amino-2-(4-hydroxyphenyl-2,3,5,6-d4)acetic-2-d acid, (2 R)-2-Amino-2-(4-hydroxyphenyl-2,3,5,6-d4)acetic-2-d acid, (2 R)-2-Amino-2-(4-hydroxyphenyl-2,3,5,6-d4)acetic-2-d acid |
| InChI Formula | InChI=1 S/C8H9NO3/c9-7(8(11)12)5-1-3-6(10)4-2-5/h1-4,7,10 H,9H2,(H,11,12)/t7-/m1/s1/i1D,2 D,3 D,4 D,7 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1O)[C@@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 R)-2-amino-2-deuterio-2-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetic acid |
| Molecular Weight (g/mol) | 172.1928 |
| Formula Weight | 172.0896 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction