Learn More
Cytidine-5,6-d2, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Cytidine-5,6-D2 |
| CAS | 40632-25-5 |
| Molecular Formula | C9 2H2 H11 N3 O5 |
| Quantity | 0.005 g |
| Synonym | Cytidine-5,6-d2, 5,6-Dideuteriocytidine, 1-β-D-Ribofuranosylcytosine-4,5-d2, 4-Amino-1-(β-D-ribofuranosyl)-2(1 H)-pyrimidinone-4,5-d2, [4,5-2H2]Cytidine, Cytidine-5,6-D2, Cytidine-d2 |
| InChI Formula | InChI=1 S/C9H13N3O5/c10-5-1-2-12(9(16)11-5)8-7(15)6(14)4(3-13)17-8/h1-2,4,6-8,13-15 H,3H2,(H2,10,11,16)/t4-,6-,7-,8-/m1/s1/i1D,2 D |
| SMILES | [2 H]C1=C([2 H])C(=NC(=O)N1[C@@H]2 O[C@H](CO)[C@@H](O)[C@H]2 O)N |
| IUPAC Name | 4-amino-5,6-dideuterio-1-[(2 R,3 R,4 S,5 R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
| Molecular Weight (g/mol) | 245.23 |
| Formula Weight | 245.0981 g/mol |
| Show More |