missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophosphamide-d8 H2O [bis(2-chloroethyl-d4)], CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Cyclophosphamide-d8 H2O [bis(2-chloroethyl-d4)] |
| Molecular Formula | C7H9D8Cl2N2O3P |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2/i2D2,3D2,5D2,6D2; |
| SMILES | O=P1(N(C([2 H])([2 H])C([2 H])([2 H])Cl)C([2 H])([2 H])C([2 H])([2 H])Cl)NCCCO1.O |
| IUPAC Name | 2-(bis(2-chloroethyl-1,1,2,2-d4)amino)-1,3,2-oxazaphosphinane 2-oxide hydrate |
| Molecular Weight (g/mol) | 287.15 |
| Formula Weight | 286.09 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction