Learn More
Coumarin-5,6,7,8-d4, CDN
Product Code. 30321869
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321869
Brand: CDN CDND55860.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Coumarin5,6,7,8-D4
CAS 185056-83-1
Molecular Formula C9 2H4 H2 O2
Quantity 0.01 g
Synonym 2 H-1-Benzopyran-2-one-5,6,7,8-d4 (9 CI), Coumarin 5,6,7,8-D4, Coumarin-d4
InChI Formula InChI=1 S/C9H6O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6 H/i1D,2 D,3 D,4 D
SMILES [2 H]c1c([2 H])c([2 H])c2C=CC(=O)Oc2c1[2 H]
IUPAC Name 5,6,7,8-tetradeuteriochromen-2-one
Molecular Weight (g/mol) 150.17
Formula Weight 150.0619 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity