Learn More
Colchicine-d3 (N-acetyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Colchicine 7-acetamide-d3 |
| CAS | 1217625-62-1 |
| Molecular Formula | C22 2H3 H22 N O6 |
| Quantity | 0.005 g |
| Synonym | 2,2,2-Trideuterio-N-[(7 S)-1,2,3,10-tetramethoxy-9-oxo-6,7-dihydro-5 H-benzo[a]heptalen-7-yl]acetamide, 2,2,2-Trideuterio-N-[(7 S)-1,2,3,10-tetramethoxy-9-oxo-6,7-dihydro-5 H-benzo[d]heptalen-7-yl]acetamide, Colchicine-d3, Colchicine-D3 (7-acetamide-D3), N-[(7 S)-1,2,3,10-Tetramethoxy-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]acetamide-2,2,2-d3, N-[(7 S)-5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl]acetamide-2,2,2-d3 |
| InChI Formula | InChI=1 S/C22H25NO6/c1-12(24)23-16-8-6-13-10-19(27-3)21(28-4)22(29-5)20(13)14-7-9-18(26-2)17(25)11-15(14)16/h7,9-11,16 H,6,8H2,1-5H3,(H,23,24)/t16-/m0/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)N[C@H]1 CCc2cc(OC)c(OC)c(OC)c2C3=CC=C(OC)C(=O)C=C13 |
| IUPAC Name | 2,2,2-trideuterio-N-[(7 S)-1,2,3,10-tetramethoxy-9-oxo-6,7-dihydro-5 H-benzo[a]heptalen-7-yl]acetamide |
| Molecular Weight (g/mol) | 402.46 |
| Formula Weight | 402.187 g/mol |
| Show More |