Learn More
Chlorpromazine-d6 HCl (N,N-dimethyl-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Chlorpromazine-d6 Hydrochloride |
| CAS | 1228182-46-4 |
| Molecular Formula | C17 2H6 H13 Cl N2 S . Cl H |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C17H19ClN2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;/h3-4,6-9,12 H,5,10-11H2,1-2H3;1 H/i1D3,2D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(CCCN1c2ccccc2Sc3ccc(Cl)cc13)C([2 H])([2 H])[2 H] |
| IUPAC Name | 3-(2-chlorophenothiazin-10-yl)-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 361.36 |
| Formula Weight | 360.1101 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |