missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbofuran-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Carbofuran-d3 |
| CAS | 1007459-98-4 |
| Molecular Formula | C12 2H3 H12 N O3 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C12H15NO3/c1-12(2)7-8-5-4-6-9(10(8)16-12)15-11(14)13-3/h4-6 H,7H2,1-3H3,(H,13,14)/i3D3 |
| SMILES | [2 H]C([2 H])([2 H])NC(=O)Oc1cccc2CC(C)(C)Oc12 |
| IUPAC Name | (2,2-dimethyl-3 H-1-benzofuran-7-yl) N-(trideuteriomethyl)carbamate |
| Molecular Weight (g/mol) | 224.27 |
| Formula Weight | 224.124 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction