missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbamoylcholine-d9 Chloride (N,N,N-trimethyl-d9), CDN
Click to view available options
Quantity:
0.01 g
0.025 g
Unit Size:
0.01g
0.03g
Specifications
Specifications
| Chemical Name or Material | Carbachol-D9 (N,N,N-trimethyl-D9) |
| Molecular Formula | C6 D9 H6 N2 O2 . Cl |
| Quantity | 0.01 g |
| Synonym | Carbamoylcholine-d9 Chloride (N,N,N-trimethyl-d9), 2-[(Aminocarbonyl)oxy]-N,N,N-tri(methyl-d3)ethanaminium chloride, Carbamoylcholine-d9 chloride, Carbachol-d9, Carbachol-D9 (N,N,N-trimethyl-D9) |
| InChI Formula | InChI=1 S/C6H14N2O2.ClH/c1-8(2,3)4-5-10-6(7)9;/h4-5H2,1-3H3,(H-,7,9);1 H/i1D3,2D3,3D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](CCOC(=O)N)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-carbamoyloxyethyl-tris(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 191.704 |
| Formula Weight | 191.1387 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction