Learn More
Captan-3,3,4,5,6,6-d6, CDN
Product Code. 30320347
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320347
Brand: CDN CDND73400.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Captan-4,4,5,6,7,7-D6
CAS 1330190-00-5
Molecular Formula C9 2H6 H2 Cl3 N O2 S
Quantity 0.01 g
Synonym Captan-4,4,5,6,7,7-D6, Captan D6
InChI Formula InChI=1 S/C9H8Cl3NO2S/c10-9(11,12)16-13-7(14)5-3-1-2-4-6(5)8(13)15/h1-2,5-6 H,3-4H2/i1D,2 D,3D2,4D2
SMILES [2 H]C1=C([2 H])C([2 H])([2 H])C2C(C(=O)N(SC(Cl)(Cl)Cl)C2=O)C1([2 H])[2 H]
IUPAC Name 4,4,5,6,7,7-hexadeuterio-2-(trichloromethylsulfanyl)-3 A,7 A-dihydroisoindole-1,3-dione
Molecular Weight (g/mol) 306.63
Formula Weight 304.9718 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity