missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benomyl-d4 (benzimidazole-4,5,6,7-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Benomyl-d4 (benzimidazole-4,5,6,7-d4) |
| CAS | 1398065-98-9 |
| Molecular Formula | C14 D4 H14 N4 O3 |
| Quantity | 0.01 g |
| Synonym | Benomyl-d4, Benomyl-d4 (benzimidazole-4,5,6,7-d4), Benomyl-D4 (benzimidazole-4,5,6,7-D4), Methyl 1-(butylcarbamoyl)-2-benzimidazolyl-4,5,6,7-d4-carbamate, [1-[(Butylamino)carbonyl]-1 H-benzimidazol-2-yl-4,5,6,7-d4]carbamic acid methyl ester |
| InChI Formula | InChI=1 S/C14H18N4O3/c1-3-4-9-15-13(19)18-11-8-6-5-7-10(11)16-12(18)17-14(20)21-2/h5-8 H,3-4,9H2,1-2H3,(H,15,19)(H,16,17,20)/i5D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(nc(NC(=O)OC)n2C(=O)NCCCC)c1[2 H] |
| IUPAC Name | methyl N-[1-(butylcarbamoyl)-4,5,6,7-tetradeuteriobenzimidazol-2-yl]carbamate |
| Molecular Weight (g/mol) | 294.342 |
| Formula Weight | 294.163 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction